siRNA Details
| Gene Symbol: LOC100131878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125681.1 | - | 1 | 619 | ||
| Gene Symbol: NRDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017021394.1 | XP_016876883.1 | 15 | 3903 | ||

LOC100131878
Genomic Map