siRNA Details
| Gene Symbol: LOC283575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110554.1 | 4 | 646 | |||
| GenBank mRNA | AK092322.1 | - | 4 | 631 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283575| Gene Symbol: LOC283575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110554.1 | 4 | 646 | |||
| GenBank mRNA | AK092322.1 | - | 4 | 631 | ||