siRNA Details
| Gene Symbol: RRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042908.1 | - | 1 | 24 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM914492.1 | - | 1 | 22 | Chr. 10 - 97393044 - 97393987 [-] on Build GRCh38 | |

RRP12
Genomic Map