siRNA Details
| Gene Symbol: RPS11P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL137354.1 | - | 1 | 293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS11P6| Gene Symbol: RPS11P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL137354.1 | - | 1 | 293 | ||