siRNA Details
| Gene Symbol: RPS11P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL137354.1 | - | 2 | 759 | ||
| BC028883.1 | - | 3 | 541 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG677644.1 | - | 3 | 540 | Chr. 12 - 64352326 - 64396649 [+] on Build GRCh38 | |

RPS11P6
Genomic Map