siRNA Details
Gene Symbol: LINC00535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033858.1 | 11 | 1824 | |||
GenBank mRNA | BC037909.2 | - | 11 | 1824 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033858.1 | 11 | 1824 | |||
GenBank mRNA | BC037909.2 | - | 11 | 1824 |