siRNA Details
Gene Symbol: LINC00535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033858.1 | 6, 7 | 1044 | |||
GenBank mRNA | BC037909.2 | - | 6, 7 | 1044 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000501400.1 | - | 6, 7 | 1044 | Chr. 8 - 93346467 - 93700433 [-] on Build GRCh38 |
