siRNA Details
| Gene Symbol: CNTRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011518172.2 | XP_011516474.1 | 33 | 5419 | ||
| XM_017014227.1 | XP_016869716.1 | 32 | 5437 | |||
| XM_017014228.1 | XP_016869717.1 | 32 | 5293 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR601314.1 | - | 1 | 784 | Chr. 9 - 121160526 - 121161505 [+] on Build GRCh38 | |

CNTRL
Genomic Map