siRNA Details
| Gene Symbol: CNTRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011518172.2 | XP_011516474.1 | 33 | 5548 | ||
| XM_017014227.1 | XP_016869716.1 | 32 | 5566 | |||
| XM_017014228.1 | XP_016869717.1 | 32 | 5422 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNTRL| Gene Symbol: CNTRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011518172.2 | XP_011516474.1 | 33 | 5548 | ||
| XM_017014227.1 | XP_016869716.1 | 32 | 5566 | |||
| XM_017014228.1 | XP_016869717.1 | 32 | 5422 | |||