siRNA Details
| Gene Symbol: LOC100289518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026856.1 | - | 2 | 1264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100289518| Gene Symbol: LOC100289518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026856.1 | - | 2 | 1264 | ||