siRNA Details
| Gene Symbol: GLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256310.1 | NP_001243239.1 | 15 | 2244 | ||
| XM_005246467.3 | XP_005246524.1 | 15 | 2303 | |||
| GenBank mRNA | AF097492.1 | - | 15 | 1878 | ||
| AF158555.1 | - | 16 | 2224 | |||
| BX641148.1 | - | 1 | 629 | |||
| CR749593.1 | - | 8 | 2703 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB134028.1 | - | 1 | 519 | Chr. 2 - 190932528 - 190933067 [+] on Build GRCh38 | |

GLS
Genomic Map