siRNA Details
| Gene Symbol: OXCT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125115.1 | - | 1 | 2009 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA484130.1 | - | 1 | 485 | Chr. 5 - 41732147 - 41732674 [-] on Build GRCh38 | |

OXCT1
Genomic Map