siRNA Details
| Gene Symbol: LOC101928433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136240.1 | 3 | 3295 | |||
| GenBank mRNA | AK091312.1 | - | 1 | 1116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928433| Gene Symbol: LOC101928433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136240.1 | 3 | 3295 | |||
| GenBank mRNA | AK091312.1 | - | 1 | 1116 | ||