siRNA Details
| Gene Symbol: MPHOSPH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017018677.1 | XP_016874166.1 | 22 | 4305 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC036600.1 | - | 1 | 127 | Chr. 12 - 123154215 - 123156181 [-] on Build GRCh38 | |

MPHOSPH9
Genomic Map