siRNA Details
| Gene Symbol: MPHOSPH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011537740.2 | XP_011536042.1 | 24 | 3719 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC036600.1 | - | 1 | 1794 | Chr. 12 - 123154215 - 123156181 [-] on Build GRCh38 | |

MPHOSPH9
Genomic Map