siRNA Details
| Gene Symbol: TCF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056961.1 | - | 1 | 134 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA893202.1 | - | 1 | 134 | Chr. 15 - 57238103 - 57238668 [+] on Build GRCh38 | |

TCF12
Genomic Map