siRNA Details
| Gene Symbol: LOC101928994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120648.1 | 5 | 1648 | |||
| GenBank mRNA | AK055968.1 | - | 5 | 1648 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000450995.1 | - | 5 | 1648 | Chr. 10 - 69215333 - 69232490 [-] on Build GRCh38 | |

LOC101928994
Genomic Map