siRNA Details
| Gene Symbol: LOC101928994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120648.1 | 5 | 1567 | |||
| GenBank mRNA | AK055968.1 | - | 5 | 1567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928994| Gene Symbol: LOC101928994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120648.1 | 5 | 1567 | |||
| GenBank mRNA | AK055968.1 | - | 5 | 1567 | ||