siRNA Details
| Gene Symbol: RASSF8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038227.1 | 4 | 919 | |||
| NR_038228.1 | 4 | 906 | ||||
| GenBank mRNA | AK055961.1 | - | 1 | 402 | ||
| BX109355.1 | - | 3 | 697 | |||
| DA682910.1 | - | 1 | 402 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RASSF8-AS1| Gene Symbol: RASSF8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038227.1 | 4 | 919 | |||
| NR_038228.1 | 4 | 906 | ||||
| GenBank mRNA | AK055961.1 | - | 1 | 402 | ||
| BX109355.1 | - | 3 | 697 | |||
| DA682910.1 | - | 1 | 402 | |||