siRNA Details
| Gene Symbol: RNF150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005263150.4 | XP_005263207.1 | 8 | 6937 | ||
| XM_011532147.2 | XP_011530449.1 | 8 | 6400 | |||
| XM_011532148.2 | XP_011530450.1 | 8 | 6415 | |||
| XM_017008475.1 | XP_016863964.1 | 7 | 6334 | |||
| XM_017008476.1 | XP_016863965.1 | 7 | 6307 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK001013.1 | - | 1 | 1377 | Chr. 4 - 140861176 - 140864183 [-] on Build GRCh38 | |

RNF150
Genomic Map