siRNA Details
| Gene Symbol: LOC101929340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120577.1 | 1 | 132 | |||
| GenBank mRNA | AK000493.1 | - | 1 | 138 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929340| Gene Symbol: LOC101929340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120577.1 | 1 | 132 | |||
| GenBank mRNA | AK000493.1 | - | 1 | 138 | ||