siRNA Details
Gene Symbol: LOC101929340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120577.1 | 2 | 325 | |||
NR_120579.1 | 2 | 90 | ||||
GenBank mRNA | AK000493.1 | - | 2 | 331 | ||
CB997328.1 | - | 2 | 117 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000504932.2 | - | 2 | 330 | Chr. 11 - 124759129 - 124765936 [+] on Build GRCh38 |
