siRNA Details
Gene Symbol: LOC101929340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120577.1 | 2 | 480 | |||
NR_120579.1 | 2 | 245 | ||||
GenBank mRNA | AK000493.1 | - | 2 | 486 | ||
CB997328.1 | - | 2 | 272 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120577.1 | 2 | 480 | |||
NR_120579.1 | 2 | 245 | ||||
GenBank mRNA | AK000493.1 | - | 2 | 486 | ||
CB997328.1 | - | 2 | 272 |