siRNA Details
| Gene Symbol: LOC100147773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125374.1 | 1 | 237 | |||
| GenBank mRNA | AK124964.1 | - | 1 | 237 | ||
| BC009228.2 | - | 1 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100147773| Gene Symbol: LOC100147773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125374.1 | 1 | 237 | |||
| GenBank mRNA | AK124964.1 | - | 1 | 237 | ||
| BC009228.2 | - | 1 | 195 | |||