siRNA Details
| Gene Symbol: FHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005263319.3 | XP_005263376.1 | 2 | 102 | ||
| XM_006714393.3 | XP_006714456.1 | 2 | 111 | |||
| XM_011532388.1 | XP_011530690.1 | 4 | 593 | |||
| XM_011532389.1 | XP_011530691.1 | 2 | 707 | |||
| XM_017008787.1 | XP_016864276.1 | 2 | 111 | |||

FHDC1
Genomic Map