siRNA Details
| Gene Symbol: CABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005274114.3 | XP_005274171.2 | 1 | 34 | ||
| XM_011545181.2 | XP_011543483.1 | 1 | 34 | |||
| XM_011545182.2 | XP_011543484.1 | 1 | 34 | |||
| XM_011545183.2 | XP_011543485.1 | 1 | 26 | |||
| GenBank mRNA | AK311199.1 | - | 2 | 385 | ||

CABP4
Genomic Map