siRNA Details
| Gene Symbol: CABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300896.1 | NP_001287825.1 | 1 | 162 | ||
| XM_017018025.1 | XP_016873514.1 | 1 | 171 | |||
| GenBank mRNA | AK311199.1 | - | 1 | 141 | ||
| BC033167.1 | - | 1 | 162 | 4549347 | ||

CABP4
Genomic Map