siRNA Details
| Gene Symbol: LOC101929718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110139.1 | 3 | 711 | |||
| GenBank mRNA | AK026285.1 | - | 3 | 711 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY253671.1 | - | 39 | Chr. 14 - 20996247 - 20997035 [-] on Build GRCh38 | ||

LOC101929718
Genomic Map