siRNA Details
| Gene Symbol: LOC283357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK124392.1 | - | 1 | 1837 | ||
| AL833464.1 | - | 1 | 2533 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283357| Gene Symbol: LOC283357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK124392.1 | - | 1 | 1837 | ||
| AL833464.1 | - | 1 | 2533 | |||