siRNA Details
| Gene Symbol: SPATA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093385.1 | - | 5 | 849 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR595989.1 | - | 1 | 576 | Chr. 2 - 231012058 - 231014422 [+] on Build GRCh38 | |

SPATA3
Genomic Map