siRNA Details
| Gene Symbol: LMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036442.1 | 4 | 1546 | |||
| GenBank mRNA | BC036550.1 | - | 2 | 3519 | ||
| BC047239.1 | - | 3 | 3132 | |||
| BC070105.1 | - | 2 | 3017 | |||
| BC098409.1 | - | 2 | 3017 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LMF1| Gene Symbol: LMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036442.1 | 4 | 1546 | |||
| GenBank mRNA | BC036550.1 | - | 2 | 3519 | ||
| BC047239.1 | - | 3 | 3132 | |||
| BC070105.1 | - | 2 | 3017 | |||
| BC098409.1 | - | 2 | 3017 | |||