siRNA Details
Gene Symbol: LMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036442.1 | 4 | 661 | |||
GenBank mRNA | BC036550.1 | - | 2 | 2634 | ||
BC047239.1 | - | 2 | 2624 | |||
BC070105.1 | - | 2 | 2132 | |||
BC098409.1 | - | 2 | 2132 | |||
BM017667.1 | - | 4 | 648 |
