siRNA Details
| Gene Symbol: GTF3C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012204.3 | NP_036336.2 | 5 | 4641 | ||
| NR_133925.1 | 6 | 4679 | ||||
| XM_017015318.1 | XP_016870807.1 | 5 | 2424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTF3C4| Gene Symbol: GTF3C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012204.3 | NP_036336.2 | 5 | 4641 | ||
| NR_133925.1 | 6 | 4679 | ||||
| XM_017015318.1 | XP_016870807.1 | 5 | 2424 | |||