siRNA Details
| Gene Symbol: LOC100130548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135126.1 | 3 | 2015 | |||
| GenBank mRNA | AK057520.1 | - | 3 | 2015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130548| Gene Symbol: LOC100130548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135126.1 | 3 | 2015 | |||
| GenBank mRNA | AK057520.1 | - | 3 | 2015 | ||