siRNA Details
| Gene Symbol: C17orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193655.1 | NP_001180584.1 | 1 | 86 | ||
| XM_006722293.2 | XP_006722356.1 | 1 | 689 | |||
| XM_011523606.2 | XP_011521908.1 | 1 | 689 | |||
| XM_017025072.1 | XP_016880561.1 | 1 | 689 | |||
| XM_017025073.1 | XP_016880562.1 | 1 | 689 | |||
| XM_017025075.1 | XP_016880564.1 | 1 | 689 | |||
| GenBank mRNA | AK090484.1 | - | 1 | 469 | ||
| BG338078.1 | - | 1 | 87 | |||
| BX451530.2 | - | 1 | 308 | |||

C17orf62
Genomic Map