siRNA Details
| Gene Symbol: C17orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090484.1 | - | 1 | 997 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR612860.1 | - | 1 | 1579 | Chr. 17 - 82449070 - 82450691 [-] on Build GRCh38 | |

C17orf62
Genomic Map