siRNA Details
| Gene Symbol: NKTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011533747.2 | XP_011532049.1 | 8 | 1985 | ||
| XM_017006476.1 | XP_016861965.1 | 2 | 3179 | |||
| GenBank mRNA | AB209888.1 | - | 6 | 3904 | ||
| BX538094.1 | - | 3952 | ||||
| BX640751.1 | - | 1 | 1194 | |||
| BX641127.1 | - | 1 | 1495 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK093752.1 | - | 1 | 1757 | Chr. 3 - 42626924 - 42629010 [+] on Build GRCh38 | |

NKTR
Genomic Map