siRNA Details
| Gene Symbol: NKTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011533747.2 | XP_011532049.1 | 8 | 1907 | ||
| XM_017006476.1 | XP_016861965.1 | 2 | 3101 | |||
| GenBank mRNA | AB209888.1 | - | 6 | 3826 | ||
| BX538094.1 | - | 3874 | ||||
| BX640751.1 | - | 1 | 1116 | |||
| BX641127.1 | - | 1 | 1417 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK093752.1 | - | 1 | 1679 | Chr. 3 - 42626924 - 42629010 [+] on Build GRCh38 | |

NKTR
Genomic Map