siRNA Details
Gene Symbol: NKTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011533747.2 | XP_011532049.1 | 8 | 990 | ||
XM_017006476.1 | XP_016861965.1 | 2 | 2184 | |||
GenBank mRNA | AB209888.1 | - | 6 | 2909 | ||
BX538094.1 | - | 2957 | ||||
BX640751.1 | - | 1 | 199 | |||
BX641127.1 | - | 1 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK093752.1 | - | 1 | 762 | Chr. 3 - 42626924 - 42629010 [+] on Build GRCh38 |
