siRNA Details
| Gene Symbol: N4BP2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020342.1 | XP_016875831.1 | 7 | 3758 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK094967.1 | - | 1 | 2313 | Chr. 13 - 32510518 - 32513484 [-] on Build GRCh38 | |

N4BP2L2
Genomic Map