siRNA Details
| Gene Symbol: CD44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF086543.1 | - | 1 | 143 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | W96291.1 | - | 1 | 143 | Chr. 11 - 35212550 - 35213708 [+] on Build GRCh38 | |

CD44
Genomic Map