siRNA Details
Gene Symbol: WDR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511469.2 | XP_011509771.1 | 12 | 5396 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF086549.1 | - | 1 | 164 | Chr. 2 - 202877109 - 202877526 [-] on Build GRCh38 |
