siRNA Details
| Gene Symbol: LOC100131599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126221.1 | - | 1 | 2411 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100131599| Gene Symbol: LOC100131599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126221.1 | - | 1 | 2411 | ||