siRNA Details
| Gene Symbol: NHSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530934.2 | XP_011529236.1 | 8 | 17830 | ||
| XM_017029473.1 | XP_016884962.1 | 8 | 16800 | |||
| XM_017029474.1 | XP_016884963.1 | 8 | 12942 | |||
| XM_017029475.1 | XP_016884964.1 | 4 | 16185 | |||
| XM_017029476.1 | XP_016884965.1 | 4 | 12302 | |||
| XM_017029477.1 | XP_016884966.1 | 4 | 17216 | |||
| GenBank mRNA | AK054940.1 | - | 1 | 1297 | ||
| AK056217.1 | - | 1 | 2047 | |||

NHSL2
Genomic Map