siRNA Details
Gene Symbol: LOC286083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111948.1 | 1 | 1178 | |||
GenBank mRNA | BC030648.2 | - | 1 | 1179 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000617093.1 | - | 1178 | Chr. - - [-] on Build |
