siRNA Details
| Gene Symbol: SPTLC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004863.3 | NP_004854.1 | 12 | 7546 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPTLC2| Gene Symbol: SPTLC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004863.3 | NP_004854.1 | 12 | 7546 | ||