siRNA Details
Gene Symbol: KCNK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022358.3 | NP_071753.2 | 2 | 1449 | ||
GenBank mRNA | BC082981.1 | - | 1 | 237 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KCNK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022358.3 | NP_071753.2 | 2 | 1449 | ||
GenBank mRNA | BC082981.1 | - | 1 | 237 |