siRNA Details
| Gene Symbol: KCNK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022358.3 | NP_071753.2 | 2 | 1288 | ||
| GenBank mRNA | BC082981.1 | - | 1 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK15| Gene Symbol: KCNK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022358.3 | NP_071753.2 | 2 | 1288 | ||
| GenBank mRNA | BC082981.1 | - | 1 | 76 | ||