siRNA Details
| Gene Symbol: KCNK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022358.3 | NP_071753.2 | 2 | 1866 | ||
| GenBank mRNA | BC082981.1 | - | 1 | 654 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM049330.1 | - | 1 | 655 | Chr. 20 - 44750995 - 44751718 [+] on Build GRCh38 | |

KCNK15
Genomic Map