siRNA Details
| Gene Symbol: CADM3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037870.1 | 4 | 2586 | |||
| GenBank mRNA | AL050219.1 | - | 1 | 311 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CADM3-AS1| Gene Symbol: CADM3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037870.1 | 4 | 2586 | |||
| GenBank mRNA | AL050219.1 | - | 1 | 311 | ||